missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferrous Sulfate, Heptahydrate, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical
F1060, 7782-63-0, FeSO4·7H2O
Supplier: Spectrum Chemical Mfg Cor F106025KG
Description
Spectrum™ Chemical Ferrous Sulfate, Heptahydrate, Crystal, Reagent, ACS As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.Specifications
| 7782-63-0 | |
| 2.5 kg | |
| MFCD00149719 | |
| O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O | |
| 278.01 | |
| ACS |
| 100% | |
| FeH14O11S | |
| SURQXAFEQWPFPV-UHFFFAOYSA-L | |
| λ²-iron(2+) heptahydrate sulfate | |
| 99% | |
| Amber Glass Bottle |
Chemical Identifiers
| 7782-63-0 | |
| 278.01 | |
| SURQXAFEQWPFPV-UHFFFAOYSA-L | |
| O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O |
| FeH14O11S | |
| MFCD00149719 | |
| λ²-iron(2+) heptahydrate sulfate |