missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Sulfate, Heptahydrate, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical

Catalog No. 18602711
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
6 x 125 g
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602711 125 g Amber Glass Bottle
18602713 500 g Amber Glass Bottle
18602712 6 x 125 g Amber Glass Bottle
18602715 2.5 kg Amber Glass Bottle
18602714 6 x 500 g Amber Glass Bottle
18602716 4 x 2.5 kg Amber Glass Bottle
18602717 12 kg Poly Pail
18602718 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18602711 Supplier Spectrum Chemical Mfg Cor Supplier No. F1060125GM

F1060, 7782-63-0, FeSO4·7H2O

Spectrum™ Chemical Ferrous Sulfate, Heptahydrate, Crystal, Reagent, ACS As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 7782-63-0
Molecular Formula FeH14O11S
Molecular Weight (g/mol) 278.01
MDL Number MFCD00149719
InChI Key SURQXAFEQWPFPV-UHFFFAOYSA-L
IUPAC Name λ²-iron(2+) heptahydrate sulfate
SMILES O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

Specifications

CAS Max % 100%
Quantity 125 g
Percent Purity 99%
Grade ACS
Packaging Amber Glass Bottle