missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Sulfate Heptahydrate, ACS Reagent (Iron (II) Sulfate Heptahydrate), Reagents

Catalog No. 01337961
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
25 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337961 100 g
01337960 10 kg
01337959 2.5 kg
01337958 25 kg
01337957 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01337961 Supplier Labchem Inc Supplier No. C1207000100C2
Only null left

Ferrous Sulfate Heptahydrate, ACS Reagent (Iron (II) Sulfate Heptahydrate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7782-63-0
Molecular Formula FeH14O11S
Molecular Weight (g/mol) 278.01
MDL Number MFCD00149719
InChI Key SURQXAFEQWPFPV-UHFFFAOYSA-L
Synonym Iron(II) Sulfate Heptahydrate
IUPAC Name λ²-iron(2+) heptahydrate sulfate
SMILES O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

Specifications

Boiling Point >300°C
Color Green or Bluish-Green
Physical Form Solid
Quantity 100 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 1.897
Chemical Name or Material Ferrous Sulfate Heptahydrate, ACS Reagent (Iron (II) Sulfate Heptahydrate)