missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferrous Sulfate Heptahydrate, >99%, MP Biomedicals
Cell Culture Reagent
$109.54 - $109.54
Chemical Identifiers
| CAS | 7782-63-0 |
|---|---|
| Molecular Formula | FeH14O11S |
| Molecular Weight (g/mol) | 278.01 |
| MDL Number | MFCD00149719 |
| InChI Key | SURQXAFEQWPFPV-UHFFFAOYSA-L |
| Synonym | iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate |
| PubChem CID | 62662 |
| ChEBI | CHEBI:75836 |
| IUPAC Name | λ²-iron(2+) heptahydrate sulfate |
| SMILES | O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19466383
|
MP Biomedicals Inc
0219466383 |
250 g |
Each for $109.54
|
|
|||||
Chemical Identifiers
| 7782-63-0 | |
| 278.01 | |
| SURQXAFEQWPFPV-UHFFFAOYSA-L | |
| 62662 | |
| λ²-iron(2+) heptahydrate sulfate |
| FeH14O11S | |
| MFCD00149719 | |
| iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate | |
| CHEBI:75836 | |
| O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O |
Specifications
| 7782-63-0 | |
| >99% | |
| MFCD00149719 | |
| SURQXAFEQWPFPV-UHFFFAOYSA-L | |
| λ²-iron(2+) heptahydrate sulfate | |
| 62662 | |
| 278 |
| 250 g | |
| FeH14O11S | |
| iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate | |
| O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O | |
| 278.01 | |
| CHEBI:75836 | |
| Ferrous Sulfate Heptahydrate |
Safety and Handling
Recommended Storage : Room Temperature