missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferrous Sulfate Heptahydrate, >99%, MP Biomedicals
Specifications
Specifications
| Physical Form | Granular Powder |
| Quantity | 250 g |
| Assay Percent Range | >99% |
| Formula Weight | 278 |
| Percent Purity | ≥99% |
| Grade | Not Available |
| Chemical Name or Material | Ferrous Sulfate Heptahydrate |
Safety and Handling
| Recommended Storage | Room Temperature |