missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Sulfate Heptahydrate, >99%, (Crystalline), MP Biomedicals

Catalog No. ICN15804883
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15804883 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15804883 Supplier MP Biomedicals Inc Supplier No. 0215804883

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity crystalline powder intended for laboratory research applications.

Chemical Identifiers

CAS 7782-63-0
Molecular Formula FeH14O11S
Molecular Weight (g/mol) 278.01
MDL Number MFCD00149719
InChI Key SURQXAFEQWPFPV-UHFFFAOYSA-L
Synonym iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
PubChem CID 62662
ChEBI CHEBI:75836
IUPAC Name λ²-iron(2+) heptahydrate sulfate
SMILES O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

Specifications

Physical Form Crystalline
Quantity 250 g
Assay Percent Range >99%
Formula Weight 278
Percent Purity 99%
Chemical Name or Material Ferrous Sulfate Heptahydrate
Recommended Storage Room Temperature