missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Gluconate, USP, 97-102%, Spectrum™ Chemical

Catalog No. 18602703
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602703 500 g Poly Bottle
18602704 2.5 kg Poly Pail
18602705 12 kg Poly Pail
18602706 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18602703 Supplier Spectrum Chemical Mfg Cor Supplier No. FE109500GMPL
Only null left

FE109, 12389-15-0, C12H22FeO14·2H2O

Spectrum™ Chemical Ferrous Gluconate, USP is used to treat iron deficiency anemia. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 12389-15-0
Molecular Formula C12H26FeO16
Molecular Weight (g/mol) 482.17
InChI Key OKGNXSFAYMSVNN-UHFFFAOYNA-L
IUPAC Name λ²-iron(2+) bis(2,3,4,5,6-pentahydroxyhexanoate) dihydrate
SMILES O.O.[Fe++].OCC(O)C(O)C(O)C(O)C([O-])=O.OCC(O)C(O)C(O)C(O)C([O-])=O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 97 to 102%
Grade USP
Loss on Drying 6.5-10%
Packaging Poly Bottle