missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Ammonium Sulfate Solution, Certified, 0.497N ±0.010N, LabChem™

Catalog No. LC145452
Change view
Click to view available options
Quantity:
1 L
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC145452 1 L Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC145452 Supplier LabChem Supplier No. LC145452
Only null left

Chemical Identifiers

CAS 7783-85-9 , 7732-18-5 , 7664-93-9
Molecular Formula FeH20N2O14S2
Molecular Weight (g/mol) 392.13
MDL Number MFCD00150530
InChI Key MQLVWQSVRZVNIP-UHFFFAOYSA-L
Synonym Ammonium Iron (II) Sulfate, Iron (II) Ammonium Sulfate Hexahydrate, Iron(II) ammonium sulfate
PubChem CID 115148  
IUPAC Name λ²-iron(2+) diammonium hexahydrate disulfate
SMILES [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Color Green
CAS Max % 19.5, 79.5, 1
Physical Form Liquid
Quantity 1 L
Linear Formula Fe(NH4)2(SO4)2·6H2O
UN Number UN3264
Solubility Information Soluble in water
Formula Weight 392.14
Grade Certified
Concentration or Composition (by Analyte or Components) 0.497N ±0.010N
Identification Passes Test
Packaging Amber Glass
Decomposition Information Gaseous Ammonia; Sulfur compounds; Thermal decomposition generates: Corrosive vapors
Chemical Name or Material Ferrous Ammonium Sulfate Solution
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature