missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Ammonium Sulfate, 0.1N, Standardized Solution, Spectrum™ Chemical

Catalog No. 18610553
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610553 4 L Amber Glass Bottle
18610552 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18610553 Supplier Spectrum Chemical Mfg Cor Supplier No. F1624LTGL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Ferrous Ammonium Sulfate
Color light green
CAS 7783-85-9, 7732-18-5, 7664-93-9
Concentration 0.1 N ± 0.006 N
Grade Chemical Solution
Packaging Amber Glass Bottle
pH Acidic
Quantity 4 L

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.