missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferrous Ammonium Sulfate, 0.0500 N (N/20), Ricca Chemical
$144.91 - $144.91
Chemical Identifiers
| CAS | 7783-85-9 |
|---|---|
| Molecular Formula | FeH20N2O14S2 |
| Molecular Weight (g/mol) | 392.13 |
| MDL Number | MFCD00150530 |
| InChI Key | MQLVWQSVRZVNIP-UHFFFAOYSA-L |
| Synonym | Ammonium Iron (II) Sulfate, Iron (II) Ammonium Sulfate Hexahydrate, Iron(II) ammonium sulfate |
| PubChem CID | 115148 |
| IUPAC Name | λ2-iron(2+) diammonium hexahydrate disulfate |
| SMILES | [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
Chemical Identifiers
| 7783-85-9 | |
| 392.13 | |
| MQLVWQSVRZVNIP-UHFFFAOYSA-L | |
| 115148 | |
| [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
| FeH20N2O14S2 | |
| MFCD00150530 | |
| Ammonium Iron (II) Sulfate, Iron (II) Ammonium Sulfate Hexahydrate, Iron(II) ammonium sulfate | |
| λ2-iron(2+) diammonium hexahydrate disulfate |
Specifications
| Green | |
| 1.82,93.0,3.41 | |
| FeH20N2O14S2 | |
| Ammonium Iron (II) Sulfate, Iron (II) Ammonium Sulfate Hexahydrate, Iron(II) ammonium sulfate | |
| [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O | |
| 392.13 | |
| Laboratory | |
| 500 mL | |
| 0.0500Normal |
| 7783-85-9,7732-18-5,7664-93-9 | |
| 1.89,97.0,3.55 | |
| MFCD00150530 | |
| MQLVWQSVRZVNIP-UHFFFAOYSA-L | |
| λ2-iron(2+) diammonium hexahydrate disulfate | |
| 115148 | |
| Amber Glass Bottle | |
| Liquid | |
| Ferrous Ammonium Sulfate |