missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Ammonium Sulfate, 0.00282 N (0.00282 M), 1 mL = 0.1 mg Cl2, Ricca Chemical

Catalog No. R3144000100
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R3144000100 100 mL Amber Glass Bottle
314416 500 mL Amber Glass Bottle
314432 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R3144000100 Supplier Ricca Chemical Company Supplier No. R3144000100C

Chemical Identifiers

CAS 7783-85-9
Molecular Formula FeH20N2O14S2
Molecular Weight (g/mol) 392.13
MDL Number MFCD00150530
InChI Key MQLVWQSVRZVNIP-UHFFFAOYSA-L
IUPAC Name λ2-iron(2+) diammonium hexahydrate disulfate
SMILES [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Color Green
Equivalence 1 mL = 0.1 mg Cl2
Grade Laboratory
Packaging Amber Glass Bottle
Quantity 100 mL
Physical Form Liquid
Concentration or Composition Notes 0.00282N,0.00282M
Chemical Name or Material Ferrous Ammonium Sulfate