missing translation for 'onlineSavingsMsg'
Learn More

Ferrosoferric Oxide, Black, Powder, Spectrum™ Chemical

Catalog No. p-7157425
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602687 500 g Amber Glass Bottle
18602688 2.5 kg Amber Glass Bottle
18602689 12 kg Poly Pail
18602690 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18602687 Supplier Spectrum Chemical Mfg Cor Supplier No. F1058500GM
Only null left

F1058, 1317-61-9, Fe3O4

Spectrum™ Chemical Ferrosoferric Oxide, Black, Powder

Chemical Identifiers

CAS 1317-61-9
Molecular Formula Fe3O4
Molecular Weight (g/mol) 231.53
MDL Number MFCD00011010
InChI Key WTFXARWRTYJXII-UHFFFAOYSA-N
IUPAC Name diiron(3+) λ²-iron(2+) tetraoxidandiide
SMILES [O--].[O--].[O--].[O--].[Fe++].[Fe+3].[Fe+3]

Specifications

CAS Max % 100%
Quantity 500 g
Grade Reagent
Packaging Amber Glass Bottle