missing translation for 'onlineSavingsMsg'
Learn More

Ferrosoferric Oxide, Black, Powder, Spectrum™ Chemical

Catalog No. 18602690
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602690 45 kg Amber Glass Bottle
18602687 500 g Amber Glass Bottle
18602688 2.5 kg Amber Glass Bottle
18602689 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18602690 Supplier Spectrum Chemical Mfg Cor Supplier No. F105845KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

F1058, 1317-61-9, Fe3O4

Spectrum™ Chemical Ferrosoferric Oxide, Black, Powder

Chemical Identifiers

CAS 1317-61-9
Molecular Formula Fe3O4
Molecular Weight (g/mol) 231.53
MDL Number MFCD00011010
InChI Key WTFXARWRTYJXII-UHFFFAOYSA-N
IUPAC Name diiron(3+) λ2-iron(2+) tetraoxidandiide
SMILES [O--].[O--].[O--].[O--].[Fe++].[Fe+3].[Fe+3]

Specifications

CAS Max % 100%
Physical Form Crystalline Powder or Needles
Quantity 45 kg
Grade Ungraded
Packaging Amber Glass Bottle
Chemical Name or Material Ferrosoferric Oxide, Black, Powder