missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferroin Indicator Solution, 0.025M, Honeywell Fluka™
Redox Indicator, 0.025 M
$70.26
Chemical Identifiers
| CAS | 14634-91-4 |
|---|---|
| Molecular Formula | C36H24FeN6O4S |
| Molecular Weight (g/mol) | 692.53 |
| MDL Number | MFCD00036428 |
| InChI Key | CIWXFRVOSDNDJZ-UHFFFAOYSA-L |
| PubChem CID | 84567 |
| IUPAC Name | iron(2+);1,10-phenanthroline;sulfate |
| SMILES | [Fe++].[O-]S([O-])(=O)=O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1 |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 14634-91-4 | |
| 692.53 | |
| CIWXFRVOSDNDJZ-UHFFFAOYSA-L | |
| iron(2+);1,10-phenanthroline;sulfate |
| C36H24FeN6O4S | |
| MFCD00036428 | |
| 84567 | |
| [Fe++].[O-]S([O-])(=O)=O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1 |
Specifications
| 0°C | |
| 14634-91-4 | |
| [Fe(C12H8N2)3]SO4 | |
| NONH for all modes of transport | |
| [Fe++].[O-]S([O-])(=O)=O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1 | |
| 692.53 | |
| 692.52g/mol | |
| 1.006g/mL at 20°C | |
| Liquid |
| 100°C | |
| C36H24FeN6O4S | |
| MFCD00036428 | |
| CIWXFRVOSDNDJZ-UHFFFAOYSA-L | |
| iron(2+);1,10-phenanthroline;sulfate | |
| 84567 | |
| Odorless | |
| Red | |
| Ferroin indicator solution |