missing translation for 'onlineSavingsMsg'
Learn More

Ferroin Indicator Solution, 0.025M, Solstice

Catalog No. 60046934
Change view
Click to view available options
Quantity:
50 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046934 50 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60046934 Supplier Solstice Supplier No. 4627050ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Redox Indicator, 0.025 M

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 14634-91-4
Molecular Formula C36H24FeN6O4S
Molecular Weight (g/mol) 692.53
MDL Number MFCD00036428
InChI Key CIWXFRVOSDNDJZ-UHFFFAOYSA-L
PubChem CID 84567
IUPAC Name iron(2+);1,10-phenanthroline;sulfate
SMILES [Fe++].[O-]S([O-])(=O)=O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1

Specifications

Melting Point 0°C
Boiling Point 100°C
Odor Odorless
Packaging Glass Bottle
Linear Formula [Fe(C12H8N2)3]SO4
UN Number NONH for all modes of transport
Density 1.006g/mL at 20°C
Color Red
Quantity 50 mL
Physical Form Liquid
Formula Weight 692.52g/mol
Chemical Name or Material Ferroin indicator solution
Show More Show Less