missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ferritin Light Chain Monoclonal Antibody (H1)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA566063
Description
Ferritin Light Chain Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes the light subunit of the ferritin protein. Ferritin is the major intracellular iron storage protein in prokaryotes and eukaryotes. It is composed of 24 subunits of the heavy and light ferritin chains. Variation in ferritin subunit composition may affect the rates of iron uptake and release in different tissues. A major function of ferritin is the storage of iron in a soluble and nontoxic state. Defects in this light chain ferritin gene are associated with several neurodegenerative diseases and hyperferritinemia-cataract syndrome. This gene has multiple pseudogenes.Specifications
| Ferritin Light Chain | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P02792, P02793, P19133 | |
| FTL, Ftl1 | |
| Recombinant Ferritin, Light Polypeptide (FTL) (Ser2-Asp175). | |
| 200 μL | |
| Primary | |
| Human, Rat, Pig | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| H1 | |
| Unconjugated | |
| FTL | |
| ferritin L subunit; Ferritin L subunit 1; ferritin L-chain; ferritin light chain; Ferritin light chain 1; ferritin light polypeptide-like 3; ferritin, light polypeptide; fril; Ftl; Ftl1; Ftl-1; L-ferritin; LFTD; NBIA3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2512, 29292, 397035 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |