missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferric Sulfate, Powder, 72%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor F1045125GM
Specifications
| 15244-10-7 | |
| 125 g | |
| VXWSFRMTBJZULV-UHFFFAOYSA-H | |
| diiron(3+) hydrate trisulfate | |
| 72% | |
| Amber Glass Bottle |
| 1 | |
| Fe2H2O13S3 | |
| O.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O | |
| 417.87 | |
| Reagent |
Chemical Identifiers
| 15244-10-7 | |
| 417.87 | |
| diiron(3+) hydrate trisulfate |
| Fe2H2O13S3 | |
| VXWSFRMTBJZULV-UHFFFAOYSA-H | |
| O.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.