missing translation for 'onlineSavingsMsg'
Learn More

Ferric Sulfate, Powder, 72%, Spectrum™ Chemical

Catalog No. p-7165551
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610543 25 g Amber Glass Bottle
18610544 125 g Amber Glass Bottle
18610546 500 g Amber Glass Bottle
18610548 2.5 kg Poly Pail
18610545 12 kg Poly Pail
18610547 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18610543 Supplier Spectrum Chemical Mfg Cor Supplier No. F104525GM
Only null left

Chemical Identifiers

CAS 15244-10-7
Molecular Formula Fe2H2O13S3
Molecular Weight (g/mol) 417.87
InChI Key VXWSFRMTBJZULV-UHFFFAOYSA-H
IUPAC Name diiron(3+) hydrate trisulfate
SMILES O.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 72%
Grade Reagent
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.