missing translation for 'onlineSavingsMsg'
Learn More

Ferric Sulfate, MP Biomedicals

Catalog No. p-3547113
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15804290 500 g
ICN15804291 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15804290 Supplier MP Biomedicals Inc Supplier No. 0215804290
Only null left

Chemical Identifiers

CAS 10028-22-5
Molecular Formula Fe2O12S3
Molecular Weight (g/mol) 399.858
InChI Key RUTXIHLAWFEWGM-UHFFFAOYSA-H
Synonym ferric sulfate, iron iii sulfate, iron persulfate, iron tersulfate, diiron tris sulphate, diiron trisulfate, ferric persulfate, ferric tersulfate, iron sesquisulfate, ferric sesquisulfate
PubChem CID 24826
ChEBI CHEBI:53438
IUPAC Name iron(3+);trisulfate
SMILES [O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3].[Fe+3]

Specifications

Quantity 500 g
Formula Weight 399.9
Chemical Name or Material Ferric Sulfate
Recommended Storage Room Temperature