accessibility menu, dialog, popup

UserName

FeFeOSOOOOSOOOOSOOO

CAS RN 10028-22-5

IUPAC Name: diiron(3+) trisulfate
Synonyms: ferric sulfate iron tersulfate iron(3+) sulfate ferric tersulfate diiron trisulfate diiron tris(sulphate) iron persulfate iron(III) sulfate ferric sesquisulfate iron sesquisulfate
Molecular Weight (g/mol): 399.86
Molecular Formula: Fe2O12S3
InChi Key: RUTXIHLAWFEWGM-UHFFFAOYSA-H
SMILES: [Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results