missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medisca Ferric Subsulfate USP (Monsels Solution)
Description
Ferric Subsulfate USP is a reddish brown liquid, odorless or nearly so. It is affected by light.
Alternative name(s): ferric subsulfate solution
Specifications
Specifications
| Chemical Name or Material | Ferric Subsulfate USP (Monsels Solution) |
| Packaging | Bottle, 120mL, Amber Glass, Boston Round, 22-400 |
| CAS | 1310-45-8 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 737.704 |
| Quantity | 100 mL |
| Percent Purity | 20 to 22 g of iron per 100 mL |
| Grade | USP |