missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferric Pyrophosphate, Powder, FCC, 24-26%, Spectrum™ Chemical
F1036, 10058-44-3, Fe4(P2O7)3·xH2O
$742.75 - $6843.18
Chemical Identifiers
| CAS | 10058-44-3 |
|---|---|
| Molecular Formula | Fe4O21P6 |
| Molecular Weight (g/mol) | 745.20 |
| MDL Number | MFCD00016091 |
| InChI Key | CADNYOZXMIKYPR-UHFFFAOYSA-B |
| IUPAC Name | tetrairon(3+) tri(phosphonatooxy)phosphonate |
| SMILES | [Fe+3].[Fe+3].[Fe+3].[Fe+3].[O-]P([O-])(=O)OP([O-])([O-])=O.[O-]P([O-])(=O)OP([O-])([O-])=O.[O-]P([O-])(=O)OP([O-])([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18602676
|
Spectrum Chemical Mfg Cor
F1036500GM |
500 g | Poly Pail |
Each for $742.75
|
|
||||
|
18602677
|
Spectrum Chemical Mfg Cor
F103625KG |
2.5 kg | Poly Pail |
Each for $2,984.93
|
|
||||
|
18602678
|
Spectrum Chemical Mfg Cor
F103612KG |
12 kg | Fiber Drum |
Each for $3,812.71
|
|
||||
|
18602679
|
Spectrum Chemical Mfg Cor
F103625KGBL |
25 kg | Fiber Drum |
Each for $6,843.18
|
|
||||
Description
Spectrum™ Chemical Ferric Pyrophosphate, Powder, FCC is used as an iron fortification food supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 10058-44-3 | |
| 745.20 | |
| CADNYOZXMIKYPR-UHFFFAOYSA-B | |
| [Fe+3].[Fe+3].[Fe+3].[Fe+3].[O-]P([O-])(=O)OP([O-])([O-])=O.[O-]P([O-])(=O)OP([O-])([O-])=O.[O-]P([O-])(=O)OP([O-])([O-])=O |
| Fe4O21P6 | |
| MFCD00016091 | |
| tetrairon(3+) tri(phosphonatooxy)phosphonate |
Specifications
| 10058-44-3 | |
| 500 g | |
| MFCD00016091 | |
| [Fe+3].[Fe+3].[Fe+3].[Fe+3].[O-]P([O-])(=O)OP([O-])([O-])=O.[O-]P([O-])(=O)OP([O-])([O-])=O.[O-]P([O-])(=O)OP([O-])([O-])=O | |
| 745.20 | |
| FCC | |
| Poly Pail |
| 100% | |
| Fe4O21P6 | |
| CADNYOZXMIKYPR-UHFFFAOYSA-B | |
| tetrairon(3+) tri(phosphonatooxy)phosphonate | |
| 24 to 26% | |
| 20% |