missing translation for 'onlineSavingsMsg'
Learn More

Ferric Pyrophosphate, Powder, FCC, 24-26%, Spectrum™ Chemical

Catalog No. 18602676
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
25 kg
Packaging:
Fiber Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602676 500 g Poly Pail
18602677 2.5 kg Poly Pail
18602678 12 kg Fiber Drum
18602679 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18602676 Supplier Spectrum Chemical Mfg Cor Supplier No. F1036500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

F1036, 10058-44-3, Fe4(P2O7)3·xH2O

Spectrum™ Chemical Ferric Pyrophosphate, Powder, FCC is used as an iron fortification food supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 10058-44-3
Molecular Formula Fe4O21P6
Molecular Weight (g/mol) 745.20
MDL Number MFCD00016091
InChI Key CADNYOZXMIKYPR-UHFFFAOYSA-B
IUPAC Name tetrairon(3+) tri(phosphonatooxy)phosphonate
SMILES [Fe+3].[Fe+3].[Fe+3].[Fe+3].[O-]P([O-])(=O)OP([O-])([O-])=O.[O-]P([O-])(=O)OP([O-])([O-])=O.[O-]P([O-])(=O)OP([O-])([O-])=O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 24 to 26%
Grade FCC
Loss on Ignition 20%
Packaging Poly Pail