missing translation for 'onlineSavingsMsg'
Learn More

Ferric Oxide, Red, Powder, Technical, Spectrum™ Chemical

Catalog No. 18602672
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602672 500 g Amber Glass Bottle
18602673 2.5 kg Amber Glass Bottle
18602674 12 kg Amber Glass Bottle
18602675 45 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18602672 Supplier Spectrum Chemical Mfg Cor Supplier No. F1040500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

F1040, 1309-37-1, Fe2O3

Spectrum™ Chemical Ferric Oxide, Red, Powder, Technical , also known as iron(II) oxide, is one of the main oxides of iron. It is sometimes referred to as rust and also occurs naturally as the mineral magnetite. Technical grade products supplied by Spectrum are indicative of a grade suitable for general industrial use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 1309-37-1
Molecular Formula Fe2O3
Molecular Weight (g/mol) 159.69
MDL Number MFCD00011008
InChI Key JEIPFZHSYJVQDO-UHFFFAOYSA-N
IUPAC Name trioxodiiron
SMILES [Fe][Fe](=O)(=O)=O

Specifications

CAS Max % 100%
Physical Form Powder, Needles or Crystals
Quantity 500 g
Grade Technical
Packaging Amber Glass Bottle
Chemical Name or Material Ferric Oxide, Red, Powder