missing translation for 'onlineSavingsMsg'
Learn More

Ferric Nitrate, Nonahydrate, Crystal, Technical, Spectrum™ Chemical

$381.53 - $6,723.47

Chemical Identifiers

CAS 7782-61-8
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18610528
View Documents
Spectrum Chemical Mfg Cor
F1031500GM
500 g Poly Bottle N/A
Only null left
 
18610529
View Documents
Spectrum Chemical Mfg Cor
F103125KG
2.5 kg Poly Bottle N/A
Only null left
 
18610530
View Documents
Spectrum Chemical Mfg Cor
F103112KG
12 kg Poly Pail N/A
Only null left
 
18610531
View Documents
Spectrum Chemical Mfg Cor
F103150KGBL
50 kg Fiber Drum N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7782-61-8

Specifications

7782-61-8
500 g
Poly Bottle
1
Technical
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.