missing translation for 'onlineSavingsMsg'
Learn More

Ferric Nitrate, Nonahydrate, Crystal, Technical, Spectrum™ Chemical

Catalog No. 18610531
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610531 50 kg Poly Bottle
18610528 500 g Poly Bottle
18610529 2.5 kg Poly Bottle
18610530 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18610531 Supplier Spectrum Chemical Mfg Cor Supplier No. F103150KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7782-61-8 | Weight: 404.00 g/mol

Chemical Identifiers

CAS 7782-61-8

Specifications

Physical Form Crystals and/or Chunks
CAS Max % 1
Quantity 50 kg
Packaging Poly Bottle
Molecular Weight (g/mol) 404.00
Grade Technical
Chemical Name or Material Ferric Nitrate, Nonahydrate, Crystal

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.