missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferric Citrate, Purified, Spectrum™ Chemical
F1028, 2338-05-8, Approx. FeC6H5O7·xH2O
$361.80 - $18482.90
Chemical Identifiers
| CAS | 2338-05-8 |
|---|---|
| Molecular Formula | C6H5FeO7 |
| Molecular Weight (g/mol) | 244.94 |
| InChI Key | NPFOYSMITVOQOS-UHFFFAOYSA-K |
| IUPAC Name | iron(3+) 2-hydroxypropane-1,2,3-tricarboxylate |
| SMILES | [Fe+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18602667
|
Spectrum Chemical Mfg Cor
F1028125GM |
125 g | Poly Bottle |
Each for $361.80
|
|
||||
|
18602668
|
Spectrum Chemical Mfg Cor
F1028500GM |
500 g | Poly Bottle |
Each for $966.07
|
|
||||
|
18602669
|
Spectrum Chemical Mfg Cor
F102825KG |
2.5 kg | Poly Pail |
Each for $3,264.40
|
|
||||
|
18602670
|
Spectrum Chemical Mfg Cor
F102812KG |
12 kg | Poly Pail |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18602671
|
Spectrum Chemical Mfg Cor
F102850KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Ferric Citrate, Purified is a powder with a green to reddish-brown appearance. It can also be used in water purification, printing and an acidity regulator.Chemical Identifiers
| 2338-05-8 | |
| 244.94 | |
| iron(3+) 2-hydroxypropane-1,2,3-tricarboxylate |
| C6H5FeO7 | |
| NPFOYSMITVOQOS-UHFFFAOYSA-K | |
| [Fe+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O |
Specifications
| 2338-05-8 | |
| 125 g | |
| NPFOYSMITVOQOS-UHFFFAOYSA-K | |
| iron(3+) 2-hydroxypropane-1,2,3-tricarboxylate | |
| Purified |
| 100% | |
| C6H5FeO7 | |
| [Fe+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O | |
| 244.94 | |
| Poly Bottle |