missing translation for 'onlineSavingsMsg'
Learn More

Ferric Citrate, Purified, Spectrum™ Chemical

Catalog No. 18602671
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602671 50 kg Fiber Drum
18602667 125 g Poly Bottle
18602668 500 g Poly Bottle
18602669 2.5 kg Poly Pail
18602670 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18602671 Supplier Spectrum Chemical Mfg Cor Supplier No. F102850KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

F1028, 2338-05-8, Approx. FeC6H5O7·xH2O

Spectrum™ Chemical Ferric Citrate, Purified is a powder with a green to reddish-brown appearance. It can also be used in water purification, printing and an acidity regulator.

Chemical Identifiers

CAS 2338-05-8
Molecular Formula C6H5FeO7
Molecular Weight (g/mol) 244.94
InChI Key NPFOYSMITVOQOS-UHFFFAOYSA-K
IUPAC Name iron(3+) 2-hydroxypropane-1,2,3-tricarboxylate
SMILES [Fe+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

CAS Max % 100%
Quantity 50 kg
Grade Purified
Packaging Fiber Drum