missing translation for 'onlineSavingsMsg'
Learn More

Ferric Citrate, n-Hydrate, BAKER™, J.T. Baker™

Catalog No. 02002863
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02002863 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02002863 Supplier Avantor Supplier No. M37607

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A grade of chemicals for laboratory use

Chemical Identifiers

CAS 2362-10-9
Molecular Formula C6H16Cl2OSi2
Molecular Weight (g/mol) 231.26
InChI Key NBGGEWGFZUDQKZ-UHFFFAOYSA-N
Synonym 1,3-bis chloromethyl-1,1,3,3-tetramethyldisiloxane, 1,3-bis chloromethyl tetramethyldisiloxane, bis chloromethyl tetramethyldisiloxane, disiloxane, 1,3-bis chloromethyl-1,1,3,3-tetramethyl, tetramethylbis chloromethyl disiloxane, tetramethyl-1,3-bis chloromethyl disiloxane, chloromethyl chloromethyl dimethylsilyl oxy dimethylsilane, disiloxane,3-bis chloromethyl-1,1,3,3-tetramethyl, pubchem13666, acmc-2097an
PubChem CID 75385
IUPAC Name chloromethyl-[chloromethyl(dimethyl)silyl]oxy-dimethylsilane
SMILES C[Si](C)(CCl)O[Si](C)(C)CCl

Specifications

Quantity 500 g
Packaging Poly Bottle
Chemical Name or Material Ferric Citrate, n-Hydrate