accessibility menu, dialog, popup

UserName

CH3SiCH3ClOSiCH3H3CCl

CAS RN 2362-10-9

IUPAC Name: (chloromethyl)({[(chloromethyl)dimethylsilyl]oxy})dimethylsilane
Synonyms: (chloromethyl)({[(chloromethyl)dimethylsilyl]oxy})dimethylsilane
Molecular Weight (g/mol): 231.26
Molecular Formula: C6H16Cl2OSi2
InChi Key: NBGGEWGFZUDQKZ-UHFFFAOYSA-N
SMILES: C[Si](C)(CCl)O[Si](C)(C)CCl
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results