missing translation for 'onlineSavingsMsg'
Learn More

Ferric Chloride TS, USP Test Solution, Spectrum™ Chemical

Catalog No. p-7165579
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18610516 100 mL
18610517 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18610516 Supplier Spectrum Chemical Mfg Cor Supplier No. F166100ML
Only null left

Specifications

Chemical Name or Material Ferric Chloride TS
Color Orange
CAS 10025-77-1, 7732-18-5
CAS Max % 9, 91
Grade USP
Packaging Amber Glass Bottle
Quantity 100 mL
Physical Form Chemical Solution

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.