missing translation for 'onlineSavingsMsg'
Learn More

Ferric Chloride Hexahydrate MP Biomedicals

Catalog No. ICN15350090
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15350090 500 g
ICN15350080 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15350090 Supplier MP Biomedicals Inc Supplier No. 0215350090

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10025-77-1
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
Synonym Chlorure Perrique, Iron Trichloride, Perchlorure de Fer
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Quantity 500 g
Formula Weight 270.3
Chemical Name or Material Ferric Chloride, Hexahydrate