missing translation for 'onlineSavingsMsg'
Learn More

Ferric Chloride Hexahydrate (Lumps/Certified ACS), Fisher Chemical™

Catalog No. I88100
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
I88100 100 g Poly Bottle
I88500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I88100 Supplier Thermo Fisher Scientific Supplier No. I88 100
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10025-77-1
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
Synonym Iron (III) Chloride
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Boiling Point 280°C
Melting Point 37°C
Color Yellow
pH 2
Physical Form Solid
Quantity 100 g
Assay Percent Range 97 to 102%
UN Number 1773
Formula Weight 270.3
Percent Purity 97 to 102%
Grade Certified ACS
Identification Pass test
Packaging Poly Bottle
Vapor Pressure Negligible
Density 1.82g/mL
Assay 97.0-102.0%
Chemical Name or Material Ferric Chloride Hexahydrate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Causes burns by all exposure routes. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:3
Flammability:0
Instability:1
DOTInformation DOT Class 8, : Corrosive