missing translation for 'onlineSavingsMsg'
Learn More

Ferric Chloride Hexahydrate, ACS Reagent (Iron (III) Chloride Hexahydrate), Reagents

Catalog No. 01337979
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
10 kg
12 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337979 10 kg
01337978 125 g
01337977 12 kg
01337976 2.5 kg
01337975 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01337979 Supplier Labchem Inc Supplier No. C120400010KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ferric Chloride Hexahydrate, ACS Reagent (Iron (III) Chloride Hexahydrate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10025-77-1
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
Synonym Iron(III) Chloride Hexahydrate
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Boiling Point 280 to 285°C
Melting Point 37°C
Color Brownish-Orange
pH Acidic
Physical Form Solid
Quantity 10 kg
Grade ACS
Packaging Poly Pail
Specific Gravity 1.82
Chemical Name or Material Ferric Chloride Hexahydrate, ACS Reagent (Iron (III) Chloride Hexahydrate)