missing translation for 'onlineSavingsMsg'
Learn More

Ferric Chloride Hexahydrate, >97%, (ACS Reagent Grade), MP Biomedicals

Catalog No. p-3547100
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19404580 100 g
ICN19404590 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19404580 Supplier MP Biomedicals Inc Supplier No. 0219404580
Only null left

Chemical Identifiers

CAS 10025-77-1
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Quantity 100 g
Assay Percent Range >97%
Formula Weight 270.3
Grade ACS Reagent
Chemical Name or Material Ferric Chloride Hexahydrate
Recommended Storage Room Temperature