missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferric Chloride Hexahydrate, >97%, (ACS Reagent Grade), MP Biomedicals
Specifications
Specifications
| Quantity | 100 g |
| Assay Percent Range | >97% |
| Formula Weight | 270.3 |
| Grade | ACS Reagent |
| Chemical Name or Material | Ferric Chloride Hexahydrate |
Safety and Handling
| Recommended Storage | Room Temperature |