missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferric Ammonium Sulfate, Saturated, Certified, LabChem™
$68.46 - $273.93
Chemical Identifiers
| CAS | 7783-83-7 |
|---|---|
| Molecular Formula | FeH28NO20S2 |
| Molecular Weight (g/mol) | 482.176 |
| InChI Key | LCPUDZUWZDSKMX-UHFFFAOYSA-K |
| Synonym | ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g |
| PubChem CID | 197097 |
| IUPAC Name | azanium;iron(3+);disulfate;dodecahydrate |
| SMILES | [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3] |
Chemical Identifiers
| 7783-83-7 | |
| 482.176 | |
| ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g | |
| azanium;iron(3+);disulfate;dodecahydrate |
| FeH28NO20S2 | |
| LCPUDZUWZDSKMX-UHFFFAOYSA-K | |
| 197097 | |
| [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3] |
Specifications
| 7783-83-7,7732-18-5 | |
| 73,27 | |
| 125 mL | |
| FeNH4S2O8·12H2O | |
| Soluble in water | |
| [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3] | |
| 482.176 | |
| 482.19 | |
| Passes Test | |
| Sulfur compounds; Gaseous Ammonia; Nitrogen oxides |
| Brown-Yellow | |
| Liquid | |
| FeH28NO20S2 | |
| ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g | |
| LCPUDZUWZDSKMX-UHFFFAOYSA-K | |
| azanium;iron(3+);disulfate;dodecahydrate | |
| 197097 | |
| Certified | |
| Poly Bottle | |
| Ferric Ammonium Sulfate, Saturated |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Warning
Recommended Storage : Room Temperature