missing translation for 'onlineSavingsMsg'
Learn More

Ferric Ammonium Sulfate Dodecahydrate (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. I753
Change view
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
I753 3 kg Glass Bottle
I75500 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I753 Supplier Thermo Fisher Scientific Supplier No. I75 3
Only null left

Chemical Identifiers

CAS 7783-83-7
Molecular Formula FeH28NO20S2
Molecular Weight (g/mol) 482.176
MDL Number MFCD00150004
InChI Key LCPUDZUWZDSKMX-UHFFFAOYSA-K
Synonym ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g
PubChem CID 197097
IUPAC Name azanium;iron(3+);disulfate;dodecahydrate
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]

Specifications

Boiling Point 230°C
Melting Point 39°C
Color Purple
pH 2.5
Physical Form Solid
Quantity 3 kg
Assay Percent Range 98.5 to 102.0%
Linear Formula FeNH4(SO4)2·12H2O
Formula Weight 482.2
Percent Purity 98.5 to 102.0%
Grade Certified ACS
Identification Pass test
Packaging Glass Bottle
Density 1.710g/cm³
Chemical Name or Material Ferric Ammonium Sulfate Dodecahydrate
Show More Show Less