missing translation for 'onlineSavingsMsg'
Learn More

Ferric Ammonium Sulfate, Dodecahydrate, ACS Grade, 98.5% to 102.0%, LabChem™

Catalog No. LC142601
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC142601 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC142601 Supplier LabChem Supplier No. LC142601

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7783-83-7
Molecular Formula FeH28NO20S2
Molecular Weight (g/mol) 482.176
InChI Key LCPUDZUWZDSKMX-UHFFFAOYSA-K
Synonym ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g
PubChem CID 197097
IUPAC Name azanium;iron(3+);disulfate;dodecahydrate
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]

Specifications

Melting Point 39°C
Color Violet
CAS Max % 100
Physical Form Solid
Quantity 500 g
Linear Formula FeNH4S2O8·12H2O
Solubility Information Soluble in water
Formula Weight 482.19
Percent Purity 98.5 to 102.0%
Grade ACS
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1710kg/m3
Decomposition Information Nitrogen oxides; Gaseous Ammonia; Sulfur compounds
Density 1,710kg/m3
Chemical Name or Material Ferric Ammonium Sulfate, Dodecahydrate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
EINECSNumber 233-382-4
Recommended Storage Room Temperature