missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pyrophosphatase, inorganic (0.1 U/μL)

Catalog No. FEREF0221
Change view
Click to view available options
Quantity:
10 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEREF0221 10 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEREF0221 Supplier Thermo Scientific™ Supplier No. EF0221

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Catalyze the hydrolysis of inorganic pyrophosphate to two orthophosphates.

Thermo Scientific Pyrophosphatase, Inorganic catalyzes the hydrolysis of inorganic pyrophosphate into two orthophosphates (see Figure 1 in Supporting data). The enzyme requires a divalent metal cation with Mg2+ conferring the highest activity.

Highlights

  • Active in Thermo Scientific buffers for DNA polymerases, RNA polymerases, and reverse transcriptases 

Specifications

Chemical Name or Material Pyrophosphatase
Concentration 0.1 U/μL
Quantity 10 U

For Research Use Only. Not for use in diagnostic procedures.