Learn More
Thermo Scientific™ XceI (NspI) (10 U/μL)
Cut at RCATG^Y sites with XceI (NspI) restriction enzyme, which performs best at 37°C in Tango buffer (Isoschizomers: BstNSI, NspI).
Supplier: Thermo Scientific™ ER1471
Description
Lambda DNA digested with XceI (NspI), 0.7% agarose, 32 cleavage sites
conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
5'...R C A T GâµY...3'
3'...YâµG T A C R...5'
Conditions for 100% Activity
- 1X Buffer Tango: 33mM Tris-acetate (pH 7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate and 0.1mg/mL BSA
- Incubate at 37°C
Storage Buffer
- XceI is supplied in:10mM Tris-HCl (pH 7.5 at 25°C), 300mM KCl, 1mM DTT, 0.1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol
Ligation and Recleavage
- After 50-fold overdigestion with XceI, more than 95% of the DNA fragments can be ligated and recut
Methylation Effects
- Dam: never overlaps-no effect
- Dcm: never overlaps-no effect
- CpG: may overlap-no effect
- EcoKI: never overlaps-no effect
- EcoBI: never overlaps-no effect
Compatible Ends
- PaeI, Hin1II
Specifications
10 U/μL | |
XceI (NspI) | |
Yes | |
No | |
Restriction Enzyme |
Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive | |
10X Buffer Tango | |
37°C | |
500 U | |
Traditional Cloning |
For Research Use Only. Not for use in diagnostic procedures.