missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TopVision Agarose

Catalog No. p-4529628
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERR0491 100 g
FERR0492 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERR0491 Supplier Thermo Scientific™ Supplier No. R0491
Only null left

TopVision Agarose is highly purified agarose suitable for analytical and preparative electrophoresis of nucleic acids.

Thermo Scientific TopVision Agarose provides optimal concentration between 0.7 to 2% in all typical buffer systems. It is a highly purified agarose with very low EEO values certified by strict quality control test procedures.

Specifications

Separation Range 100 to >30000 bp
Melting Point 88 °C
Quantity 100 g
Validated Application Gel Electrophoresis
Product Line TopVision
Product Type Agarose Gel

For Research Use Only. Not for use in diagnostic procedures.