missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SspI (10 U/μL)

The SspI restriction enzyme recognizes AAT^ATT sites and cuts best at 37°C in G buffer.

$90.18 - $326.87

Specifications

Concentration 10 U/μL
Methylation Sensitivity Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
Enzyme Ssp I
Compatible Buffer 10X Buffer G
Sensitive to Heat Inactivation Yes
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
FERER0771
View Documents
Thermo Scientific™
ER0771
500 units N/A
Only null left
 
FERER0772
View Documents
Thermo Scientific™
ER0772
2,500 units N/A
Only null left
 
Description

Description

Lambda DNA digested with SspI, 1% agarose, 20 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

Highlights

  • Superior quality -stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities
Specifications

Specifications

10 U/μL
Ssp I
Yes
No
Traditional Cloning
Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
10X Buffer G
37°C
Restriction Enzyme
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.