Learn More
Thermo Scientific™ ScaI (10 U/μL)
Description
Lambda DNA digested with ScaI, 0.7% agarose, 5 cleavage sites
conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
3'...T C A▵T G A...5'
Conditions for 100% Activity
- 1X Buffer ScaI:10mM Bis-Tris Propane-HCl (pH6.5 at 37°C), 10mM MgCl2, 100mM KCl and 0.1mg/mL BSA
- Incubate at 37°C
Storage Buffer
- ScaI is supplied in:10mM Tris-HCl (pH7.5 at 25°C), 50mM KCl, 1mM DTT, 0.1mM EDTA, 0.5mg/mL BSA and 50% (v/v) glycerol
Ligation and Recleavage
- After 50-fold overdigestion with ScaI, more than 80% of the DNA fragments can be ligated and more than 90% of these can be recut
Methylation Effects
- Dam: never overlaps — no effect
- Dcm: never overlaps — no effect
- CpG: never overlaps — no effect
- EcoKI: never overlaps — no effect
- EcoBI: may overlap — blocked
Digestion of Agarose-embedded DNA
- Minimum 20units of the enzyme are required for complete digestion of 1μg of agarose-embedded lambda DNA in 16hours
Note:
Conditions of low salt, high enzyme concentration, glycerol concentration > 5% or pH> 8.0 may result in star activity. Supercoiled plasmids may require up to 20-fold more ScaI for complete digestion than linear DNA.
Specifications
Specifications
| Concentration | 10 U/μL |
| Methylation Sensitivity | Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive |
| Enzyme | Sca I |
| Compatible Buffer | Unique Buffer (10X Buffer ScaI) |
| Sensitive to Heat Inactivation | Yes |
| Optimal Reaction Temperature | 37°C |
| Type IIS RE | No |
| Quantity | 1,000 units |
| Product Type | Restriction Enzyme |
| Research Category | Traditional Cloning |
Frequently Asked Questions (FAQs)
For optimal results with fast reaction and 100% buffer compatibility, we highly recommend using FastDigest restriction enzymes in double digestion. In certain cases however, it may be possible to perform double digestion using a mix of Thermo Scientific conventional and Fastdigest restriction enzymes. For specific recommendations, please contact our technical service with detailed information about the enzymes and DNA template you plan to use.
We recommend only 2 µl 10X Buffer in digestion of unpurified PCR products in 30 ul since salts and ions from the PCR reaction would be carried over to the digestion reaction.
Star activty may be contributed by:
• Prolonged incubation
• High enzyme concentration
• High glycerol concentration (usually 5% or higher)
• Small reaction volume
Unexpected cleavage patterns may be caused by the following reasons:
• Star activity of the restriction enzyme: Make sure to follow the reaction recommendations as specified in the protocol. Star activity may be improved by changing several key factors such as decreasing the reaction time, increasing the reaction volume, and decreasing the enzyme amount.
• Partial or incomplete cleavage (incomplete restriction reaction): Efficiency of the enzyme can be improved by adding more enzyme, prolonging the reaction time, or purifying DNA samples to remove inhibitory contaminants.
• Contamination with non-specific endonucleases: Non-specific endonucleases may be introduced to the DNA sample and/or the enzyme from improper handling, pipetting, etc.
•Improper reaction setup: Mix the digestion reaction thoroughly.
The main reason for DNA cleavage reaction failure is the presence of contaminating inhibitors in the template DNA (for example: phenol, chloroform, detergents, ethanol, excess salts, EDTA, etc.). The best way to troubleshoot is to perform control reactions:
1) negative control (experimental DNA in the reaction buffer without the restriction enzyme) to access degradation of DNA by contaminants in the DNA template and/or reaction buffer
2) positive control reaction I (digestion of highly pure control DNA with the restriction enzyme) to access reaction conditions and enzyme activity
3) positive control reaction II (highly pure control DNA + experimental DNA + Restriction Enzyme) to access possible issues with the experimental DNA.
In addition, please check for sensitivity of the restriction enzymes to template DNA methylation.
For Research Use Only. Not for use in diagnostic procedures.