missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PstI (10 U/μL)

The PstI restriction enzyme recognizes CTGCA^G sites and cuts best at 37°C in O buffer (Isoschizomers: BspMAI).

Supplier:  Thermo Scientific™ ER0612

 View more versions of this product

Catalog No. FERER0612


Only null left

Description

Description

Lambda DNA digested with PstI, 0.7% agarose, 28 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'..C T G C A▵G...3'

3'..G▵A C G T C...5'

Conditions for 100% Activity

  • 1X Buffer O:50mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2, 100mM NaCl and 0.1mg/mL BSA
  • Incubate at 37°C

Storage Buffer

  • PstI is supplied in: 10mM Tris-HCl (pH7.4 at 25°C), 200mM NaCl, 1mM DTT, 0.1mM EDTA, 0.15% Triton X-100, 0.2mg/mL BSA and 50% (v/v) glycerol

Ligation and Recleavage

  • After 50-fold overdigestion with PstI, more than 95% of the DNA fragments can be ligated and recut

Methylation Effects

  • Dam: never overlaps —no effect
  • Dcm: never overlaps —no effect
  • CpG: never overlaps —no effect
  • EcoKI: never overlaps —no effect
  • EcoBI: never overlaps —no effect

Digestion of Agarose-embedded DNA

  • Minimum 5units of the enzyme are required for complete digestion of 1μg of agarose-embedded lambda DNA in 16hours

Compatible Ends

  • Alw21I, ApaI, BseSI, Eco24I, Mph1103I, PstI, SacI, SdaI

Note:

Conditions of high pH, low salt, high glycerol, 8% DMSO can cause star activity (Malyguine, E., et al., Gene, 8, 163-177, 1980). Surrounding sequences: the presence of adjacent runs of G-C base pairs confers significant resistance to cleavage (Armstrong, K. and Bauer, W.R., NAR, 10, 993-1007, 1982). 100% dUTP incorporation at the recognition site reduces PstI cleavage to 25% (Glenn, T.C., et al., Biotechniques, 17, 1086-1090, 1994). PstI will not cut AGCTGCAG when methylated by AluI methyltransferase.

Specifications

Chemical Identifiers

10 U/μL

Specifications

10 U/μL
Pst I
No
No
Restriction Enzyme
Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
10X Buffer O
37°C
5 x 3,000 units
Traditional Cloning
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.