Learn More
Thermo Scientific™ PscI (PciI) (10 U/μL)
The PscI (PciI) restriction enzyme recognizes A^CATGT sites and cuts best at 37°C in Tango buffer (Isoschizomers: PciI).
Supplier: Thermo Scientific™ ER1871
Description
Lambda DNA digested with PscI (PciI), 0.7% agarose, 2 cleavage sites
conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
5'...AâµC A T G T...3'
3'...T G T A CâµA...5'
Conditions for 100% Activity
- 1X Buffer Tango:33mM Tris-acetate (pH7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate and 0.1mg/mL BSA
- Incubate at 37°C
Storage Buffer
- PscI is supplied in:10mM Tris-HCl (pH7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol
Ligation and Recleavage
- After 50-fold overdigestion with PscI, more than 95% of the DNA fragments can be ligated and recut
Methylation Effects
- Dam: never overlaps —no effect
- Dcm: never overlaps —no effect
- CpG: never overlaps —no effect
- EcoKI: may overlap —effect not determined
- EcoBI: may overlap —effect not determined
Digestion of Agarose-embedded DNA
- Minimum 5units of the enzyme are required for complete digestion of 1μg of agarose-embedded lambda DNA in 16hours
Compatible Ends
- AflIII, BtgI, Eco130I, FatI, NcoI, PagI
Note:
PscI, like its isoschizomers BspLU11I and PciI, is inhibited by nonionic detergents Triton X-100 (>0.002%) and Nonidet P40 (>0.001%).
Specifications
| 10 U/μL | |
| PscI (PciI) | |
| Yes | |
| No | |
| Restriction Enzyme |
| Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive | |
| 10X Buffer Tango | |
| 37°C | |
| 200 units | |
| Traditional Cloning |
For Research Use Only. Not for use in diagnostic procedures.