missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ pBR322 Plasmid DNA

Catalog No. FERSD0041
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERSD0041 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERSD0041 Supplier Thermo Scientific™ Supplier No. SD0041
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Optimize cloning with Thermo Scientific™ pBR322 DNA, a 4361 bp double-stranded closed circular medium copy plasmid DNA.

pBR322 is one of the most commonly used E.coli cloning vectors.

Highlights

  • Isolated from E. coli (dam+, dcm+)
  • More than 90% in the supercoiled form
  • Purified by chromatography using proprietary patented technology
  • For DNA sequence, sequence analysis, and map creation, see the REviewer online free tool.

Specifications

Cloning Method Restriction Enzyme⁄ MCS
Concentration 0.5 μg/μL
Sample Type DNA
Final Product Type DNA
For Use With (Application) Cloning
Product Type Plasmid DNA
Quantity 100 μg
Vector pBR322

For Research Use Only. Not for use in diagnostic procedures.