missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ NTP Set, 100 mM Solution
Description
The Thermo Scientific NTP set is a convenient set of 100 mM solutions of each ATP, CTP, GTP and UTP. These highly stable nucleotides are more than 99% pure and can be used in a variety of molecular biology applications.
Features
• Greater than 99% purity confirmed by HPLC
• Functionally tested in in vitro transcription
• Stable for two years at -20°C and after multiple freeze-thaw cycles
Applications
• aRNA synthesis
• siRNA synthesis
• RNA amplification
• in vitro transcription
Related products
100 mM ATP Solution (Cat. No. R0441)
100 mM CTP Solution (R0451)
100 mM GTP Solution (R0461)
100 mM UTP Solution (R0471)
Specifications
Specifications
| Concentration | 100 mM |
| For Use With (Application) | in vitro Transcription, RNA Amplification |
| Content And Storage | NTP Set, 100 mM solution (4 x 0.25 mL) Store at –20°C. |
| Shipping Condition | Dry Ice |
| Conjugate | Unconjugated |
| Quantity | 4 x 0.25 mL |
| Product Type | NTP Set |
| Form | Liquid |
For Research Use Only. Not for use in diagnostic procedures.