Learn More
Thermo Scientific™ HinfI, HC (50 U/μL)
Description
Thermo Scientific™ HinfI is available as a FastDigest enzyme which is used for rapid DNA digestion. More than 95% of the digested DNA fragments can be ligated and recut at a 5'-termini concentration of 1.3μM after a 50-fold overdigestion (3U/μg DNA x 17 hours) with HinfI.
5'...G▵A N T C...3'
3'...C T N A▵G...5' Conditions for 100% Activity:
- 1X Buffer R: 10mM Tris-HCl (pH8.5 at 37°C), 10mM MgCl2, 100mM KCl and 0.1mg/mL BSA
- Incubate at 37°C
Storage Buffer:
- HinfI is supplied in: 10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol
Methylation Effects:
- Dam: never overlaps —no effect
- Dcm: never overlaps —no effect
- CpG: may overlap —cleavage impaired
- EcoKI: never overlaps —no effect
- EcoBI: may overlap —blocked
Compatible Ends:
- G▴AWTC — PfeI
Specifications
Specifications
| Concentration | 50 U/μL |
| Methylation Sensitivity | CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive |
| Enzyme | Hinf I |
| Compatible Buffer | 10X Buffer R |
| Sensitive to Heat Inactivation | Yes |
| Optimal Reaction Temperature | 37°C |
| Type IIS RE | No |
| Quantity | 10,000 units |
| Product Type | Restriction Enzyme |
| Research Category | Traditional Cloning |
Frequently Asked Questions (FAQs)
For optimal results with fast reaction and 100% buffer compatibility, we highly recommend using FastDigest restriction enzymes in double digestion. In certain cases however, it may be possible to perform double digestion using a mix of Thermo Scientific conventional and Fastdigest restriction enzymes. For specific recommendations, please contact our technical service with detailed information about the enzymes and DNA template you plan to use.
We recommend only 2 µl 10X Buffer in digestion of unpurified PCR products in 30 ul since salts and ions from the PCR reaction would be carried over to the digestion reaction.
Star activty may be contributed by:
• Prolonged incubation
• High enzyme concentration
• High glycerol concentration (usually 5% or higher)
• Small reaction volume
Unexpected cleavage patterns may be caused by the following reasons:
• Star activity of the restriction enzyme: Make sure to follow the reaction recommendations as specified in the protocol. Star activity may be improved by changing several key factors such as decreasing the reaction time, increasing the reaction volume, and decreasing the enzyme amount.
• Partial or incomplete cleavage (incomplete restriction reaction): Efficiency of the enzyme can be improved by adding more enzyme, prolonging the reaction time, or purifying DNA samples to remove inhibitory contaminants.
• Contamination with non-specific endonucleases: Non-specific endonucleases may be introduced to the DNA sample and/or the enzyme from improper handling, pipetting, etc.
•Improper reaction setup: Mix the digestion reaction thoroughly.
The main reason for DNA cleavage reaction failure is the presence of contaminating inhibitors in the template DNA (for example: phenol, chloroform, detergents, ethanol, excess salts, EDTA, etc.). The best way to troubleshoot is to perform control reactions:
1) negative control (experimental DNA in the reaction buffer without the restriction enzyme) to access degradation of DNA by contaminants in the DNA template and/or reaction buffer
2) positive control reaction I (digestion of highly pure control DNA with the restriction enzyme) to access reaction conditions and enzyme activity
3) positive control reaction II (highly pure control DNA + experimental DNA + Restriction Enzyme) to access possible issues with the experimental DNA.
In addition, please check for sensitivity of the restriction enzymes to template DNA methylation.
For Research Use Only. Not for use in diagnostic procedures.