missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HincII (HindII) (10 U/μL)

The HincII (HindII) restriction enzyme recognizes GTY^RAC sites and cuts best at 37°C in Tango buffer (Isoschizomers: HindII).

$87.19 - $313.76

Specifications

Concentration 10 U/μL
Methylation Sensitivity CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
Enzyme HincII (HindII)
Compatible Buffer 10X Buffer Tango
Sensitive to Heat Inactivation Yes
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
FERER0491
View Documents Promotion Details
Thermo Scientific™
ER0491
500 units
Each for $87.19
Only null left
 
FERER0492
View Documents Promotion Details
Thermo Scientific™
ER0492
2,500 units
Each for $313.76
Only null left
 
Description

Description

Lambda DNA digested with HincII (HindII), 1% agarose, 35 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

Highlights

  • Superior quality -stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities
Specifications

Specifications

10 U/μL
HincII (HindII)
Yes
No
Traditional Cloning
CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
10X Buffer Tango
37°C
Restriction Enzyme
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.